Gene Transcripts

Gene Symbol
SNX10
Entrez Gene ID
29887
Gene Name
sorting nexin 10
Gene Aliases
OPTB8
Chromosome Location
Chr. 7: 26291895 - 26374330 on Build GRCh38
UniGene ID
Hs.741316
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001199835.1 NP_001186764.1 6 595
NM_001199837.1 NP_001186766.1 4 400
NM_001199838.1 NP_001186767.1 3 299
NM_001318198.1 NP_001305127.1 7 669
NM_001318199.1 NP_001305128.1 6 470
NM_013322.2 NP_037454.2 6 548
XM_006715712.2 XP_006715775.1 6 748
XM_017012085.1 XP_016867574.1 8 763
XM_017012086.1 XP_016867575.1 8 1202
XM_017012087.1 XP_016867576.1 7 692
XM_017012088.1 XP_016867577.1 7 639
GenBank mRNA AF121860.1 - 6 461
AK296142.1 - 7 499
AK307905.1 - 4 349
AK309162.1 - 3 299
AK311256.1 - 2 359
AK312850.1 - 6 470
AK316088.1 - 7 645
BC034992.1 - 6 521 4827599
BG399132.1 - 3 312
BJ995940.1 - 4 400
BX648363.1 - 1 1551
DA377368.1 - 6 470

View Full Product Details