Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_022075.4 | NP_071358.1 | 10 | 1295 | |
NM_181746.3 | NP_859530.1 | 10 | 1335 | ||
XM_011509451.2 | XP_011507753.1 | 10 | 998 | ||
XM_011509452.2 | XP_011507754.1 | 10 | 1046 | ||
GenBank mRNA | AF177338.2 | - | 10 | 1024 | |
AF189062.4 | - | 5 | 508 | ||
AK001105.1 | - | 9 | 964 | ||
AK290989.1 | - | 10 | 971 | ||
AY091458.1 | - | 10 | 1117 | ||
BC001357.2 | - | 10 | 990 | 3051086 | |
BC010032.2 | - | 10 | 954 | 3506408 |