Gene Transcripts
- Gene Symbol
- NRBP1
- Entrez Gene ID
- 29959
- Gene Name
- nuclear receptor binding protein 1
- Gene Aliases
- BCON3, MADM, MUDPNP, NRBP
- Chromosome Location
- Chr. 2: 27427790 - 27442259 on Build GRCh38
- UniGene ID
- Hs.515876
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001321357.1 |
NP_001308286.1
|
11, 12 |
1732 |
|
NM_001321358.1 |
NP_001308287.1
|
11, 12 |
1129 |
|
NM_001321359.1 |
NP_001308288.1
|
11, 12 |
1124 |
|
NM_001321362.1 |
NP_001308291.1
|
10, 11 |
1102 |
|
NM_001321363.1 |
NP_001308292.1
|
10, 11 |
1009 |
|
NM_013392.3 |
NP_037524.1
|
10, 11 |
1100 |
|
GenBank mRNA |
AF113249.1 |
- |
10, 11 |
1012 |
|
AK001946.1 |
- |
10, 11 |
990 |
|
AK027538.1 |
- |
10, 11 |
1008 |
|
AK092860.1 |
- |
3, 4 |
1410 |
|
AK122664.1 |
- |
11, 12 |
1732 |
|
AK130619.1 |
- |
3, 4 |
216 |
|
AK223136.1 |
- |
10, 11 |
996 |
|
AK225540.1 |
- |
10, 11 |
1026 |
|
AK301345.1 |
- |
11, 12 |
943 |
|
AL136682.1 |
- |
10, 11 |
987 |
|
BC001221.1 |
- |
10, 11 |
1014 |
3534543 |
View Full Product Details