Gene Transcripts

Gene Symbol
NOX3
Entrez Gene ID
50508
Gene Name
NADPH oxidase 3
Gene Aliases
GP91-3, MOX-2
Chromosome Location
Chr. 6: 155395368 - 155455903 on Build GRCh38
UniGene ID
Hs.247776
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015718.2 NP_056533.1 11 1521
GenBank mRNA AF190122.1 - 11 1521
AF229177.1 - 9 1260

View Full Product Details