Gene Transcripts

Gene Symbol
GAL
Entrez Gene ID
51083
Gene Name
galanin and GMAP prepropeptide
Gene Aliases
ETL8, GAL-GMAP, GALN, GLNN, GMAP
Chromosome Location
Chr. 11: 68684475 - 68691175 on Build GRCh38
UniGene ID
Hs.278959
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015973.4 NP_057057.2 4 421
GenBank mRNA BC030241.1 - 4 339
M77140.1 - 3 115

View Full Product Details