Gene Transcripts

Gene Symbol
HERC5
Entrez Gene ID
51191
Gene Name
HECT and RLD domain containing E3 ubiquitin protein ligase 5
Gene Aliases
CEB1, CEBP1
Chromosome Location
Chr. 4: 88456604 - 88506170 on Build GRCh38
UniGene ID
Hs.26663
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016323.3 NP_057407.2 15 2035
GenBank mRNA AB027289.1 - 15 1995
AK093155.1 - 5 1370
AK301756.1 - 5 603
AK302142.1 - 9 907
AY337518.1 - 15 2035
BC140716.1 - 15 1928 9021584

View Full Product Details