Gene Transcripts

Gene Symbol
DNAJC27
Entrez Gene ID
51277
Gene Name
DnaJ heat shock protein family (Hsp40) member C27
Gene Aliases
RBJ, RabJS
Chromosome Location
Chr. 2: 24943636 - 24972094 on Build GRCh38
UniGene ID
Hs.434993
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016544.2 NP_057628.1 6 789
XM_017004269.1 XP_016859758.1 6 820
XM_017004270.1 XP_016859759.1 6 680
GenBank mRNA AF178983.1 - 6 623
AK294726.1 - 6 573
AL137731.1 - 8 898
BC034049.1 - 6 650 4815297

View Full Product Details