Gene Transcripts

Gene Symbol
COMMD10
Entrez Gene ID
51397
Gene Name
COMM domain containing 10
Gene Aliases
PTD002
Chromosome Location
Chr. 5: 116084592 - 116446462 on Build GRCh38
UniGene ID
Hs.483136
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001308080.1 NP_001295009.1 4 391
NM_016144.3 NP_057228.1 4 364
XM_011543458.2 XP_011541760.1 4 748
XM_011543460.2 XP_011541762.1 4 747
XM_017009553.1 XP_016865042.1 4 486
XM_017009555.1 XP_016865044.1 4 746
XM_017009556.1 XP_016865045.1 4 746
GenBank mRNA AF078857.1 - 4 319
AF161423.1 - 4 315
AK002147.1 - 4 340
AY542165.1 - 4 287
BC005179.1 - 4 310 3683093
BI859871.1 - 4 380
DA001572.1 - 4 391
HY139221.1 - 4 365

View Full Product Details