Gene Transcripts

Gene Symbol
TMEM14C
Entrez Gene ID
51522
Gene Name
transmembrane protein 14C
Gene Aliases
C6orf53, HSPC194, MSTP073, NET26, bA421M1.6
Chromosome Location
Chr. 6: 10722915 - 10731129 on Build GRCh38
UniGene ID
Hs.519557
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001165258.1 NP_001158730.1 6 1092
NM_016462.3 NP_057546.1 6 1078
GenBank mRNA AF151028.1 - 6 888
AV737904.1 - 1 183
BC002496.2 - 6 813 3050609
BC010086.2 - 6 842 2907072

View Full Product Details