Gene Transcripts

Gene Symbol
YPEL5
Entrez Gene ID
51646
Gene Name
yippee like 5
Gene Aliases
CGI-127
Chromosome Location
Chr. 2: 30146568 - 30160533 on Build GRCh38
UniGene ID
Hs.515890
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001127399.1 NP_001120871.1 4 491
NM_001127400.1 NP_001120872.1 4 727
NM_001127401.1 NP_001120873.1 5 788
NM_016061.2 NP_057145.1 3 430
XM_017004318.1 XP_016859807.1 5 1026
XM_017004319.1 XP_016859808.1 4 464
XM_017004320.1 XP_016859809.1 4 965
XM_017004321.1 XP_016859810.1 4 657
XM_017004322.1 XP_016859811.1 4 888
GenBank mRNA AB098739.1 - 2 227
AF135161.1 - 3 334
AF151885.1 - 3 352
AF172940.1 - 3 354
AK025730.1 - 4 368
AK307099.1 - 2 143
AK311404.1 - 3 141
BC000836.1 - 4 305 3456031
BC047237.1 - 4 888 5269563

View Full Product Details