Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_016530.2 | NP_057614.1 | 5 | 451 | |
XM_017022312.1 | XP_016877801.1 | 6 | 506 | ||
GenBank mRNA | AB038995.1 | - | 5 | 446 | |
AK001111.1 | - | 5 | 374 | ||
AK297598.1 | - | 5 | 374 | ||
AL833365.1 | - | 5 | 364 | ||
BC020654.1 | - | 5 | 363 | 4701429 | |
BG721755.1 | - | 5 | 515 |