Gene Transcripts

Gene Symbol
TM6SF2
Entrez Gene ID
53345
Gene Name
transmembrane 6 superfamily member 2
Gene Aliases
Chromosome Location
Chr. 19: 19264365 - 19273265 on Build GRCh38
UniGene ID
Hs.531624
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001001524.2 NP_001001524.2 10 1062
GenBank mRNA AK026307.1 - 7 1346
AK097070.1 - 7 1408

View Full Product Details