Gene Transcripts
- Gene Symbol
- SH3TC1
- Entrez Gene ID
- 54436
- Gene Name
- SH3 domain and tetratricopeptide repeats 1
- Gene Aliases
- Chromosome Location
- Chr. 4: 8182072 - 8241103 on Build GRCh38
- UniGene ID
- Hs.479116
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001318480.1 |
NP_001305409.1
|
7, 8 |
1113 |
|
NM_018986.4 |
NP_061859.3
|
7, 8 |
1030 |
|
XM_006713889.2 |
XP_006713952.1
|
4, 5 |
448 |
|
XM_011513488.1 |
XP_011511790.1
|
7, 8 |
1007 |
|
XM_017008300.1 |
XP_016863789.1
|
7, 8 |
1139 |
|
XM_017008301.1 |
XP_016863790.1
|
7, 8 |
903 |
|
XM_017008302.1 |
XP_016863791.1
|
4, 5 |
448 |
|
GenBank mRNA |
AK000363.1 |
- |
3, 4 |
418 |
|
AK074402.1 |
- |
7, 8 |
907 |
|
AK126165.1 |
- |
7, 8 |
986 |
|
AK222865.1 |
- |
3, 4 |
418 |
|
View Full Product Details