Gene Transcripts

Gene Symbol
C2orf42
Entrez Gene ID
54980
Gene Name
chromosome 2 open reading frame 42
Gene Aliases
Chromosome Location
Chr. 2: 70149885 - 70191071 on Build GRCh38
UniGene ID
Hs.720049
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017880.1 NP_060350.1 9 1698
XM_005264389.2 XP_005264446.1 9 1891
XM_005264390.2 XP_005264447.1 9 1760
XM_005264391.2 XP_005264448.1 9 1842
XM_011532937.2 XP_011531239.1 8 1636
XM_017004398.1 XP_016859887.1 9 2179
XM_017004399.1 XP_016859888.1 9 1887
XM_017004400.1 XP_016859889.1 9 1771
GenBank mRNA AK000565.1 - 9 1698
AK026303.1 - 7 983
AL136834.1 - 8 1476
BC002825.2 - 9 1687 3635986
BC005079.1 - 9 1687 3615724

View Full Product Details