Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001039395.3 | NP_001034484.3 | 6 | 828 | |
XM_006716795.2 | XP_006716858.1 | 8 | 897 | ||
XM_006716798.3 | XP_006716861.1 | 9 | 902 | ||
XM_006716799.3 | XP_006716862.1 | 7 | 962 | ||
XM_006716800.2 | XP_006716863.1 | 8 | 852 | ||
XM_006716801.2 | XP_006716864.1 | 7 | 953 | ||
XM_011517952.1 | XP_011516254.1 | 8 | 840 | ||
XM_011517954.1 | XP_011516256.1 | 6 | 893 | ||
XM_011517956.1 | XP_011516258.1 | 10 | 1131 | ||
XM_011517957.1 | XP_011516259.1 | 7 | 1023 | ||
XM_011517958.1 | XP_011516260.1 | 8 | 950 | ||
XM_017014881.1 | XP_016870370.1 | 8 | 897 | ||
XM_017014882.1 | XP_016870371.1 | 8 | 897 | ||
XM_017014883.1 | XP_016870372.1 | 8 | 897 | ||
XM_017014884.1 | XP_016870373.1 | 7 | 810 | ||
XM_017014885.1 | XP_016870374.1 | 7 | 657 | ||
XM_017014886.1 | XP_016870375.1 | 6 | 709 | ||
XM_017014887.1 | XP_016870376.1 | 6 | 791 | ||
GenBank mRNA | AK000920.1 | - | 9 | 1076 | |
AK295831.1 | - | 6 | 820 | ||
AK301790.1 | - | 10 | 1231 | ||
BC034293.1 | - | 7 | 929 | 5167647 | |
BC036349.1 | - | 10 | 1000 |