Gene Transcripts

Gene Symbol
HEATR1
Entrez Gene ID
55127
Gene Name
HEAT repeat containing 1
Gene Aliases
BAP28, UTP10
Chromosome Location
Chr. 1: 236549005 - 236604541 on Build GRCh38
UniGene ID
Hs.708114
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018072.5 NP_060542.4 17 2324
GenBank mRNA AK095692.1 - 17 2287
BC130000.1 - 2 248
BC150614.1 - 16 2194 9056984

View Full Product Details