Gene Transcripts

Gene Symbol
FAM90A1
Entrez Gene ID
55138
Gene Name
family with sequence similarity 90 member A1
Gene Aliases
Chromosome Location
Chr. 12: 8221260 - 8227618 on Build GRCh38
UniGene ID
Hs.196086
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001319982.1 NP_001306911.1 6 2002
NM_018088.3 NP_060558.3 7 2159
XM_006719097.3 XP_006719160.1 3 2063
XM_011520719.1 XP_011519021.1 7 2150
XM_017019547.1 XP_016875036.1 7 2147
GenBank mRNA AK001270.1 - 6 1988
BC042608.1 - 7 2138 5167203

View Full Product Details