Gene Transcripts
- Gene Symbol
- UBA6
- Entrez Gene ID
- 55236
- Gene Name
- ubiquitin like modifier activating enzyme 6
- Gene Aliases
- E1-L2, MOP-4, UBE1L2
- Chromosome Location
- Chr. 4: 67615761 - 67701171 on Build GRCh38
- UniGene ID
- Hs.212774
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_018227.5 |
NP_060697.4
|
32, 33 |
3071 |
|
GenBank mRNA |
AB014773.1 |
- |
32, 33 |
3019 |
|
AK001670.1 |
- |
14, 15 |
1420 |
|
AK027020.1 |
- |
2, 3 |
195 |
|
AK094164.1 |
- |
24, 25 |
2277 |
|
AK314371.1 |
- |
32, 33 |
3048 |
|
AL832015.2 |
- |
32, 33 |
3040 |
|
AL832458.2 |
- |
32, 33 |
3040 |
|
AY359880.1 |
- |
19, 20 |
2392 |
|
BC126484.1 |
- |
32, 33 |
3066 |
8992200 |
BC126486.1 |
- |
32, 33 |
3066 |
8992202 |
EF623993.1 |
- |
32, 33 |
3019 |
|
View Full Product Details