Gene Transcripts

Gene Symbol
MCOLN3
Entrez Gene ID
55283
Gene Name
mucolipin 3
Gene Aliases
TRP-ML3, TRPML3
Chromosome Location
Chr. 1: 85018082 - 85048902 on Build GRCh38
UniGene ID
Hs.535239
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001253693.1 NP_001240622.1 8, 9 1082
NM_018298.10 NP_060768.8 9, 10 1250
XM_005271003.1 XP_005271060.1 10, 11 1647
XM_006710750.1 XP_006710813.1 10, 11 1466
XM_011541739.2 XP_011540041.2 9, 10 1174
XM_011541740.2 XP_011540042.1 8, 9 1008
XM_011541741.2 XP_011540043.1 10, 11 1353
XM_011541742.1 XP_011540044.1 10, 11 1384
GenBank mRNA AF475085.1 - 9, 10 1156
AK001868.1 - 8, 9 988
AK292206.1 - 9, 10 1182
AK314354.1 - 9, 10 1210

View Full Product Details