Gene Transcripts

Gene Symbol
CHRNB4
Entrez Gene ID
1143
Gene Name
cholinergic receptor nicotinic beta 4 subunit
Gene Aliases
Chromosome Location
Chr. 15: 78623282 - 78655586 on Build GRCh38
UniGene ID
Hs.624178
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000750.3 NP_000741.1 5 1428
XM_011521186.2 XP_011519488.1 7 1458
XM_011521187.2 XP_011519489.1 6 1365
XM_011521190.2 XP_011519492.1 4 1596
XM_011521191.2 XP_011519493.1 4 1176
XM_011521192.2 XP_011519494.1 5 1569
XM_017021885.1 XP_016877374.1 5 1523
XM_017021886.1 XP_016877375.1 5 3330
XM_017021887.1 XP_016877376.1 5 1332
XM_017021888.1 XP_016877377.1 5 1332
XM_017021889.1 XP_016877378.1 5 1332
GenBank mRNA BC096080.1 - 5 1431
BC096082.1 - 5 1431 40000737
U48861.1 - 5 1436
U62439.1 - 5 1402
X68275.1 - 4 1176
Y08416.1 - 5 1407

View Full Product Details