Gene Transcripts
- Gene Symbol
- TMCO6
- Entrez Gene ID
- 55374
- Gene Name
- transmembrane and coiled-coil domains 6
- Gene Aliases
- PRO1580
- Chromosome Location
- Chr. 5: 140596529 - 140647411 on Build GRCh38
- UniGene ID
- Hs.534333
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001300980.1 |
NP_001287909.1
|
9, 10 |
1222 |
|
NM_001300982.1 |
NP_001287911.1
|
7, 8 |
911 |
|
NM_018502.4 |
NP_060972.3
|
9, 10 |
1204 |
|
XM_005268477.1 |
XP_005268534.1
|
9, 10 |
1207 |
|
XM_011537663.2 |
XP_011535965.1
|
9, 10 |
1222 |
|
XM_011537665.2 |
XP_011535967.1
|
8, 9 |
1286 |
|
XM_011537668.2 |
XP_011535970.1
|
7, 8 |
959 |
|
XM_017009617.1 |
XP_016865106.1
|
9, 10 |
1204 |
|
XM_017009618.1 |
XP_016865107.1
|
9, 10 |
1222 |
|
XM_017009619.1 |
XP_016865108.1
|
8, 9 |
1010 |
|
XM_017009620.1 |
XP_016865109.1
|
8, 9 |
995 |
|
GenBank mRNA |
AK097088.1 |
- |
9, 10 |
1168 |
|
AK289495.1 |
- |
7, 8 |
869 |
|
BC001910.1 |
- |
9, 10 |
1145 |
3538001 |
BC009618.1 |
- |
4, 5 |
437 |
3901182 |
View Full Product Details