Gene Transcripts

Gene Symbol
CCR2
Entrez Gene ID
729230
Gene Name
C-C motif chemokine receptor 2
Gene Aliases
CC-CKR-2, CCR-2, CCR2A, CCR2B, CD192, CKR2, CKR2A, CKR2B, CMKBR2, MCP-1-R
Chromosome Location
Chr. 3: 46353744 - 46360940 on Build GRCh38
UniGene ID
Hs.511794
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001123041.2 NP_001116513.2 2 577
NM_001123396.1 NP_001116868.1 2 577
XM_011534069.1 XP_011532371.1 3 290
GenBank mRNA AK292685.1 - 2 587
AK292920.1 - 2 195
BC074751.2 - 1 131 30915210
BC095540.1 - 1 96 7001971
BC126452.1 - 1 126 8992168
BC136396.1 - 1 126 9020383
D29984.1 - 1 92
DC418358.1 - 2 286
U03882.1 - 1 131
U03905.1 - 2 172

View Full Product Details