Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_198795.1 | NP_942090.1 | 14 | 1880 | |
XM_005269978.2 | XP_005270035.1 | 14 | 1861 | ||
XM_005269979.2 | XP_005270036.1 | 14 | 1894 | ||
XM_011539959.2 | XP_011538261.1 | 14 | 2233 | ||
XM_011539960.2 | XP_011538262.1 | 14 | 1823 | ||
XM_011539961.2 | XP_011538263.1 | 14 | 1805 | ||
XM_011539962.1 | XP_011538264.1 | 14 | 2187 | ||
XM_011539963.1 | XP_011538265.1 | 14 | 1894 | ||
XM_017016414.1 | XP_016871903.1 | 13 | 1750 | ||
GenBank mRNA | AF285606.1 | - | 7 | 849 | |
AK024735.1 | - | 4 | 400 | ||
AK122916.1 | - | 14 | 1890 | ||
AL833340.1 | - | 14 | 1882 | ||
BC035010.2 | - | 14 | 2186 | ||
BX648664.1 | - | 14 | 1825 |