Gene Transcripts

Gene Symbol
C11orf16
Entrez Gene ID
56673
Gene Name
chromosome 11 open reading frame 16
Gene Aliases
Chromosome Location
Chr. 11: 8920076 - 8933006 on Build GRCh38
UniGene ID
Hs.277349
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020643.2 NP_065694.2 5 878
XM_017018013.1 XP_016873502.1 4 1339
GenBank mRNA AK074455.1 - 5 973
AK223377.1 - 5 973
BC027865.1 - 5 860 5203029

View Full Product Details