Gene Transcripts

Gene Symbol
C11orf16
Entrez Gene ID
56673
Gene Name
chromosome 11 open reading frame 16
Gene Aliases
Chromosome Location
Chr. 11: 8920076 - 8933006 on Build GRCh38
UniGene ID
Hs.277349
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020643.2 NP_065694.2 6 1420
XM_017018013.1 XP_016873502.1 5 1881
GenBank mRNA AK074455.1 - 6 1515
AK223377.1 - 6 1515

View Full Product Details