Gene Transcripts

Gene Symbol
MIF4GD
Entrez Gene ID
57409
Gene Name
MIF4G domain containing
Gene Aliases
AD023, MIFD, SLIP1
Chromosome Location
Chr. 17: 75266228 - 75271292 on Build GRCh38
UniGene ID
Hs.325631
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001242498.1 NP_001229427.1 4, 5 460
NM_001242500.1 NP_001229429.1 3, 4 362
NM_001242501.1 NP_001229430.1 2, 3 260
NM_020679.3 NP_065730.2 4, 5 439
XM_005257530.1 XP_005257587.1 4, 5 581
XM_005257532.4 XP_005257589.1 3, 4 337
XM_005257533.1 XP_005257590.1 3, 4 436
XM_011525051.2 XP_011523353.1 4, 5 676
XM_011525052.2 XP_011523354.1 4, 5 655
XM_011525053.2 XP_011523355.1 3, 4 553
XM_017024888.1 XP_016880377.1 4, 5 676
XM_017024889.1 XP_016880378.1 4, 5 460
XM_017024890.1 XP_016880379.1 4, 5 486
XM_017024891.1 XP_016880380.1 4, 5 398
XM_017024893.1 XP_016880382.1 3, 4 832
XM_017024894.1 XP_016880383.1 3, 4 467
GenBank mRNA AF225422.1 - 2, 3 456
AK300711.1 - 4, 5 399
BC033759.1 - 3, 4 338 5215495
BI752882.1 - 4, 5 457
BU190758.1 - 3, 4 361
CA488406.1 - 2, 3 260
KU178731.1 - 3, 4 302
KU178732.1 - 3, 4 323
KU178733.1 - 2, 3 200

View Full Product Details