Gene Transcripts

Gene Symbol
PBOV1
Entrez Gene ID
59351
Gene Name
prostate and breast cancer overexpressed 1
Gene Aliases
UC28, UROC28, dJ171N11.2
Chromosome Location
Chr. 6: 138215986 - 138218490 on Build GRCh38
UniGene ID
Hs.302016
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021635.2 NP_067648.1 1 307
GenBank mRNA AF189269.1 - 1 310
AF189270.1 - 1 310
BC069109.1 - 1 307 7216926

View Full Product Details