Gene Transcripts

Gene Symbol
AASDHPPT
Entrez Gene ID
60496
Gene Name
aminoadipate-semialdehyde dehydrogenase-phosphopantetheinyl transferase
Gene Aliases
AASD-PPT, ACPS, CGI-80, LYS2, LYS5
Chromosome Location
Chr. 11: 106077565 - 106098692 on Build GRCh38
UniGene ID
Hs.524009
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015423.2 NP_056238.2 2 447
GenBank mRNA AF136978.1 - 2 466
AF151057.1 - 2 467
AF151838.1 - 2 467
AF201943.1 - 2 416
AK293362.1 - 2 330
AK312529.1 - 2 351
BC015470.1 - 2 338 3914010
BC016728.1 - 2 367 4081713

View Full Product Details