Gene Transcripts

Gene Symbol
SPCS3
Entrez Gene ID
60559
Gene Name
signal peptidase complex subunit 3
Gene Aliases
PRO3567, SPC22/23, SPC3, YLR066W
Chromosome Location
Chr. 4: 176319939 - 176332245 on Build GRCh38
UniGene ID
Hs.42194
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021928.3 NP_068747.1 4 500
GenBank mRNA AK026302.1 - 4 475
AK092634.1 - 3 1483
AL136660.1 - 4 470
AY359044.1 - 4 458
BC047290.1 - 4 443 5224927
BC053883.1 - 3 2394
DB285025.1 - 4 502

View Full Product Details