Gene Transcripts
- Gene Symbol
- MAPKAP1
- Entrez Gene ID
- 79109
- Gene Name
- mitogen-activated protein kinase associated protein 1
- Gene Aliases
- JC310, MIP1, SIN1, SIN1b, SIN1g
- Chromosome Location
- Chr. 9: 125437394 - 125707234 on Build GRCh38
- UniGene ID
- Hs.495138
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001006617.1 |
NP_001006618.1
|
4, 5 |
839 |
|
NM_001006618.1 |
NP_001006619.1
|
4, 5 |
839 |
|
NM_001006619.1 |
NP_001006620.1
|
4, 5 |
839 |
|
NM_001006620.1 |
NP_001006621.1
|
3, 4 |
511 |
|
NM_001006621.1 |
NP_001006622.1
|
2, 3 |
421 |
|
NM_024117.3 |
NP_077022.1
|
4, 5 |
839 |
|
XM_011519005.1 |
XP_011517307.1
|
5, 6 |
980 |
|
XM_011519006.2 |
XP_011517308.1
|
5, 6 |
980 |
|
XM_011519007.1 |
XP_011517309.1
|
5, 6 |
980 |
|
XM_011519008.1 |
XP_011517310.1
|
5, 6 |
980 |
|
XM_011519009.1 |
XP_011517311.1
|
5, 6 |
980 |
|
XM_017015126.1 |
XP_016870615.1
|
4, 5 |
728 |
|
XM_017015127.1 |
XP_016870616.1
|
4, 5 |
728 |
|
GenBank mRNA |
AK290060.1 |
- |
4, 5 |
652 |
|
AK298841.1 |
- |
1, 2 |
90 |
|
AL833042.1 |
- |
3, 4 |
472 |
|
AY425957.1 |
- |
2, 3 |
209 |
|
AY524429.1 |
- |
4, 5 |
836 |
|
AY524430.1 |
- |
4, 5 |
836 |
|
AY524431.1 |
- |
4, 5 |
836 |
|
AY524432.1 |
- |
4, 5 |
836 |
|
AY633624.1 |
- |
4, 5 |
806 |
|
AY633625.1 |
- |
4, 5 |
813 |
|
AY633626.1 |
- |
4, 5 |
813 |
|
BC002326.2 |
- |
4, 5 |
808 |
2823015 |
BC003044.1 |
- |
4, 5 |
808 |
2823015 |
BX438260.2 |
- |
4, 5 |
838 |
|
M37191.1 |
- |
3, 4 |
284 |
|
View Full Product Details