Gene Transcripts

Gene Symbol
TNIP2
Entrez Gene ID
79155
Gene Name
TNFAIP3 interacting protein 2
Gene Aliases
ABIN2, FLIP1, KLIP
Chromosome Location
Chr. 4: 2741660 - 2756376 on Build GRCh38
UniGene ID
Hs.368551
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001161527.1 NP_001154999.1 6 862
NM_001292016.1 NP_001278945.1 5 899
NM_024309.3 NP_077285.3 6 1148
GenBank mRNA AF372839.1 - 6 1148
AJ304866.1 - 6 1095
AK026176.1 - 6 861
AK096296.1 - 6 1139
AL137262.1 - 1 1682
BC002740.2 - 6 1106 3632736
BX386180.2 - 5 860
CB157550.1 - 2 113

View Full Product Details