Gene Transcripts

Gene Symbol
HSPBAP1
Entrez Gene ID
79663
Gene Name
HSPB1 associated protein 1
Gene Aliases
PASS1
Chromosome Location
Chr. 3: 122739997 - 122793829 on Build GRCh38
UniGene ID
Hs.29169
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024610.5 NP_078886.2 6 915
XM_011513130.2 XP_011511432.1 7 833
XM_011513132.2 XP_011511434.2 6 674
XM_017007179.1 XP_016862668.1 5 739
XM_017007180.1 XP_016862669.1 2 377
GenBank mRNA AF400663.1 - 6 882
AK289410.1 - 6 915
BC011897.2 - 6 899 3949150

View Full Product Details