Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_024743.3 | NP_079019.3 | 1 | 456 | |
NR_024010.1 | 1 | 456 | |||
XM_011532247.2 | XP_011530549.1 | 1 | 465 | ||
GenBank mRNA | AK025587.1 | - | 1 | 261 | |
AK225308.1 | - | 1 | 261 | ||
AK293381.1 | - | 1 | 27 | ||
AY358727.1 | - | 1 | 451 | ||
AY542891.1 | - | 1 | 456 | ||
BC130533.1 | - | 1 | 449 | 40146564 |