Gene Transcripts

Gene Symbol
DHDDS
Entrez Gene ID
79947
Gene Name
dehydrodolichyl diphosphate synthase subunit
Gene Aliases
CIT, CPT, DS, HDS, RP59
Chromosome Location
Chr. 1: 26432282 - 26471306 on Build GRCh38
UniGene ID
Hs.369385
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001243564.1 NP_001230493.1 7 792
NM_001243565.1 NP_001230494.1 7 777
NM_001319959.1 NP_001306888.1 8 917
NM_024887.3 NP_079163.2 8 894
NM_205861.2 NP_995583.1 8 894
XM_006710912.2 XP_006710975.1 8 953
XM_006710913.2 XP_006710976.1 8 832
XM_006710914.2 XP_006710977.1 8 829
XM_006710915.2 XP_006710978.1 7 764
XM_006710916.2 XP_006710979.1 8 943
XM_006710917.2 XP_006710980.1 8 896
XM_006710918.2 XP_006710981.1 8 852
XM_011542183.2 XP_011540485.1 8 929
XM_011542184.2 XP_011540486.1 8 950
XM_011542185.2 XP_011540487.1 7 848
XM_011542186.2 XP_011540488.1 7 848
XM_017002379.1 XP_016857868.1 8 832
XM_017002380.1 XP_016857869.1 8 929
XM_017002381.1 XP_016857870.1 8 943
XM_017002382.1 XP_016857871.1 8 852
XM_017002383.1 XP_016857872.1 7 841
XM_017002384.1 XP_016857873.1 7 794
GenBank mRNA AB090852.1 - 7 755
AK023164.1 - 8 824
AK297134.1 - 6 712
AK308187.1 - 8 836
AK316485.1 - 7 735
BC003643.2 - 7 799 3532466
BC004117.2 - 7 719 3843062
BC034152.1 - 2 119
BX648507.1 - 8 919
GU727641.1 - 7 799

View Full Product Details