Gene Transcripts

Gene Symbol
MAP3K19
Entrez Gene ID
80122
Gene Name
mitogen-activated protein kinase kinase kinase 19
Gene Aliases
RCK, YSK4
Chromosome Location
Chr. 2: 134963875 - 135047468 on Build GRCh38
UniGene ID
Hs.659395
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001018044.2 NP_001018054.1 3 209
NM_001018046.2 NP_001018056.1 3 209
NM_001018047.2 NP_001018057.1 3 209
NM_001282883.1 NP_001269812.1 3 209
NM_001321177.1 NP_001308106.1 3 349
NM_025052.4 NP_079328.3 3 209
XM_011511891.2 XP_011510193.1 3 349
XM_011511892.1 XP_011510194.1 6 917
XM_011511894.2 XP_011510196.1 3 324
XM_011511896.2 XP_011510198.1 3 190
XM_011511897.2 XP_011510199.1 3 270
XM_017005003.1 XP_016860492.1 6 884
XM_017005004.1 XP_016860493.1 5 769
XM_017005005.1 XP_016860494.1 4 570
XM_017005006.1 XP_016860495.1 3 349
XM_017005007.1 XP_016860496.1 3 349
GenBank mRNA AK122935.1 - 3 349
AK309454.1 - 5 769
AY574901.1 - 3 209
AY574902.1 - 3 209
AY574903.1 - 3 209
AY574904.1 - 3 209
AY574905.1 - 3 209
BC137276.1 - 3 208 9021273
BC137277.1 - 3 208 9021274
BC144555.1 - 3 208

View Full Product Details