Gene Transcripts

Gene Symbol
EFHC2
Entrez Gene ID
80258
Gene Name
EF-hand domain containing 2
Gene Aliases
MRX74, dJ1158H2.1
Chromosome Location
Chr. X: 44146254 - 44343677 on Build GRCh38
UniGene ID
Hs.521953
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025184.3 NP_079460.2 15 2278
XM_006724562.2 XP_006724625.1 14 1991
GenBank mRNA AK026254.1 - 5 552
AK026496.1 - 15 2130
AK291187.1 - 15 2419
AK292967.1 - 15 2262
BC031039.1 - 15 2273 5272398
CR749363.1 - 10 1941

View Full Product Details