Gene Transcripts

Gene Symbol
C16orf70
Entrez Gene ID
80262
Gene Name
chromosome 16 open reading frame 70
Gene Aliases
C16orf6, LIN10, lin-10
Chromosome Location
Chr. 16: 67109881 - 67148544 on Build GRCh38
UniGene ID
Hs.513666
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001320540.1 NP_001307469.1 6 447
NM_001320541.1 NP_001307470.1 6 488
NM_001320542.1 NP_001307471.1 6 473
NM_001320543.1 NP_001307472.1 5 404
NM_025187.4 NP_079463.2 5 503
XM_006721284.3 XP_006721347.1 6 471
XM_011523350.2 XP_011521652.1 5 611
XM_017023729.1 XP_016879218.1 6 482
XM_017023730.1 XP_016879219.1 6 485
XM_017023731.1 XP_016879220.1 6 483
GenBank mRNA AF176088.1 - 6 403
AK022138.1 - 4 272
AL540304.3 - 6 382
BC004556.2 - 5 483 3957147
CB997191.1 - 5 362
HY157930.1 - 6 473

View Full Product Details