Gene Transcripts

Gene Symbol
ZFP91
Entrez Gene ID
80829
Gene Name
ZFP91 zinc finger protein
Gene Aliases
DMS-8, DSM-8, FKSG11, PZF, ZFP-91, ZNF757
Chromosome Location
Chr. 11: 58579114 - 58621550 on Build GRCh38
UniGene ID
Hs.524920
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001197051.1 NP_001183980.1 9 1225
NM_053023.4 NP_444251.1 9 1228
GenBank mRNA AB056107.1 - 9 1174
AF310246.1 - 9 874
AK295700.1 - 1 84
BC051743.1 - 9 1228 4827133

Gene Symbol
ZFP91-CNTF
Entrez Gene ID
386607
Gene Name
ZFP91-CNTF readthrough (NMD candidate)
Gene Aliases
Chromosome Location
Chr. 11: 58579114 - 58625733 on Build GRCh38
UniGene ID
Hs.524920
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_024091.1 9 1228
GenBank mRNA AB057443.1 - 9 1174

View Full Product Details