Gene Transcripts

Gene Symbol
HDHD3
Entrez Gene ID
81932
Gene Name
haloacid dehalogenase like hydrolase domain containing 3
Gene Aliases
2810435D12Rik, C9orf158
Chromosome Location
Chr. 9: 113373416 - 113377009 on Build GRCh38
UniGene ID
Hs.7739
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001304509.1 NP_001291438.1 3 1236
NM_001304510.1 NP_001291439.1 3 1195
NM_001304511.1 NP_001291440.1 2 1121
NM_031219.3 NP_112496.1 2 1449
GenBank mRNA AJ420424.1 - 2 952
AK315401.1 - 3 1180
BC005048.2 - 1 840 3615096
BC031878.1 - 2 1449 4747097

View Full Product Details