Gene Transcripts
- Gene Symbol
- ZMIZ2
- Entrez Gene ID
- 83637
- Gene Name
- zinc finger MIZ-type containing 2
- Gene Aliases
- NET27, TRAFIP20, ZIMP7, hZIMP7
- Chromosome Location
- Chr. 7: 44748547 - 44769881 on Build GRCh38
- UniGene ID
- Hs.600681
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001300959.1 |
NP_001287888.1
|
2, 3 |
185 |
|
NM_031449.3 |
NP_113637.3
|
2, 3 |
180 |
|
NM_174929.2 |
NP_777589.2
|
1, 2 |
119 |
|
XM_005249866.2 |
XP_005249923.1
|
2, 3 |
204 |
|
XM_005249867.4 |
XP_005249924.1
|
2, 3 |
206 |
|
XM_005249868.4 |
XP_005249925.1
|
2, 3 |
1224 |
|
XM_005249869.2 |
XP_005249926.1
|
2, 3 |
204 |
|
XM_005249870.2 |
XP_005249927.1
|
2, 3 |
204 |
|
XM_005249871.2 |
XP_005249928.1
|
2, 3 |
204 |
|
XM_005249872.2 |
XP_005249929.1
|
2, 3 |
204 |
|
XM_005249873.2 |
XP_005249930.1
|
2, 3 |
204 |
|
XM_017012674.1 |
XP_016868163.1
|
2, 3 |
285 |
|
GenBank mRNA |
AF357908.1 |
- |
1, 2 |
57 |
|
AK090415.1 |
- |
2, 3 |
180 |
|
AY426594.1 |
- |
1, 2 |
84 |
|
BC016968.1 |
- |
1, 2 |
102 |
|
BC110435.1 |
- |
2, 3 |
170 |
5518312 |
HY060154.1 |
- |
2, 3 |
185 |
|
View Full Product Details