Gene Transcripts

Gene Symbol
SYT16
Entrez Gene ID
83851
Gene Name
synaptotagmin 16
Gene Aliases
CHR14SYT, SYT14L, Strep14, syt14r, yt14r
Chromosome Location
Chr. 14: 61812156 - 62112825 on Build GRCh38
UniGene ID
Hs.404139
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_031914.2 NP_114120.2 6 1974
XM_006720272.3 XP_006720335.1 8 2434
XM_011537219.1 XP_011535521.1 8 2511
XM_011537223.2 XP_011535525.1 7 1548
XM_011537224.2 XP_011535526.1 8 1694
XM_011537225.2 XP_011535527.1 8 1993
XM_011537227.2 XP_011535529.2 6 1371
XM_011537228.2 XP_011535530.1 6 1314
XM_011537229.2 XP_011535531.1 5 1158
XM_017021693.1 XP_016877182.1 8 2511
XM_017021694.1 XP_016877183.1 8 2200
XM_017021695.1 XP_016877184.1 9 2345
XM_017021696.1 XP_016877185.1 7 1493
XM_017021698.1 XP_016877187.1 7 1617
GenBank mRNA AB102949.1 - 6 1717
AJ303367.1 - 1 128
AJ617628.1 - 6 1975
BC040924.1 - 4 866
BC117363.1 - 6 1833 40125914
BC143598.1 - 6 1833 9052110

View Full Product Details