Gene Transcripts
- Gene Symbol
- ARMC2
- Entrez Gene ID
- 84071
- Gene Name
- armadillo repeat containing 2
- Gene Aliases
- bA787I22.1
- Chromosome Location
- Chr. 6: 108848410 - 108990738 on Build GRCh38
- UniGene ID
- Hs.645481
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_032131.5 |
NP_115507.4
|
3, 4 |
463 |
|
XM_005267154.3 |
XP_005267211.1
|
3, 4 |
744 |
|
XM_005267155.2 |
XP_005267212.1
|
3, 4 |
479 |
|
XM_005267157.3 |
XP_005267214.1
|
3, 4 |
469 |
|
XM_006715573.2 |
XP_006715636.1
|
4, 5 |
569 |
|
XM_011536166.1 |
XP_011534468.1
|
4, 5 |
554 |
|
XM_011536167.1 |
XP_011534469.1
|
3, 4 |
457 |
|
XM_011536168.2 |
XP_011534470.1
|
3, 4 |
457 |
|
XM_011536170.2 |
XP_011534472.1
|
3, 4 |
457 |
|
XM_011536176.2 |
XP_011534478.1
|
3, 4 |
457 |
|
XM_017011350.1 |
XP_016866839.1
|
3, 4 |
469 |
|
XM_017011351.1 |
XP_016866840.1
|
3, 4 |
457 |
|
XM_017011352.1 |
XP_016866841.1
|
3, 4 |
469 |
|
GenBank mRNA |
AK095234.1 |
- |
2, 3 |
306 |
|
AK292499.1 |
- |
3, 4 |
457 |
|
BC030603.2 |
- |
3, 4 |
457 |
4824783 |
View Full Product Details