Gene Transcripts

Gene Symbol
C3orf20
Entrez Gene ID
84077
Gene Name
chromosome 3 open reading frame 20
Gene Aliases
Chromosome Location
Chr. 3: 14673532 - 14773036 on Build GRCh38
UniGene ID
Hs.661452
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001184957.1 NP_001171886.1 10, 11 1510
NM_001184958.1 NP_001171887.1 10, 11 1615
NM_032137.4 NP_115513.4 10, 11 2012
XM_006713341.2 XP_006713404.1 10, 11 2012
XM_006713342.2 XP_006713405.1 10, 11 1865
XM_006713343.2 XP_006713406.1 10, 11 2012
XM_011534153.2 XP_011532455.1 10, 11 3642
XM_011534154.2 XP_011532456.1 10, 11 3077
XM_011534157.2 XP_011532459.1 6, 7 789
GenBank mRNA AL136781.1 - 10, 11 1430
AL834386.1 - 10, 11 1964
BC030599.2 - 10, 11 1573 4831210
BC038406.1 - 10, 11 1932 5167835

View Full Product Details