Gene Transcripts

Gene Symbol
PCBD2
Entrez Gene ID
84105
Gene Name
pterin-4 alpha-carbinolamine dehydratase 2
Gene Aliases
DCOH2, DCOHM, PHS2
Chromosome Location
Chr. 5: 134905120 - 134962646 on Build GRCh38
UniGene ID
Hs.710014
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032151.4 NP_115527.3 3 299
GenBank mRNA AF499009.1 - 3 219
AK098495.1 - 3 271
AL136721.1 - 3 299
BC054021.1 - 3 302 6385036

View Full Product Details