Gene Transcripts

Gene Symbol
MEX3B
Entrez Gene ID
84206
Gene Name
mex-3 RNA binding family member B
Gene Aliases
MEX-3B, RKHD3, RNF195
Chromosome Location
Chr. 15: 82041778 - 82046143 on Build GRCh38
UniGene ID
Hs.104744
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032246.4 NP_115622.2 2 1138
GenBank mRNA AB095929.1 - 1 1950
AK094145.1 - 1 371
AK131424.1 - 2 1015
AY950678.1 - 2 700
BC036211.1 - 2 1136 5266259
BC111545.1 - 2 1114 5742971

View Full Product Details