Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001128855.2 | NP_001122327.1 | 5 | 706 | |
NM_001195422.1 | NP_001182351.1 | 5 | 699 | ||
NM_032620.3 | NP_116009.2 | 5 | 706 | ||
NM_133644.3 | NP_598399.2 | 5 | 706 | ||
GenBank mRNA | AF360742.1 | - | 5 | 668 | |
AF360743.1 | - | 5 | 668 | ||
AF360744.1 | - | 5 | 648 | ||
AK027606.1 | - | 5 | 653 | ||
AK091035.1 | - | 5 | 1411 | ||
AK291929.1 | - | 5 | 650 | ||
AK297953.1 | - | 5 | 681 | ||
AK298485.1 | - | 929 | |||
AK311442.1 | - | 5 | 769 | ||
AY078987.1 | - | 5 | 668 | ||
AY078988.1 | - | 5 | 668 | ||
BC008924.2 | - | 3 | 246 | ||
BC017207.2 | - | 5 | 638 | 3634066 | |
BC019261.1 | - | 5 | 647 | 3501739 | |
BM789389.1 | - | 2 | 170 | ||
DA490131.1 | - | 1 | 214 |