Gene Transcripts
- Gene Symbol
- GPAT3
- Entrez Gene ID
- 84803
- Gene Name
- glycerol-3-phosphate acyltransferase 3
- Gene Aliases
- AGPAT 10, AGPAT10, AGPAT8, AGPAT9, HMFN0839, LPAAT-theta, MAG1
- Chromosome Location
- Chr. 4: 83535634 - 83605875 on Build GRCh38
- UniGene ID
- Hs.99196
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001256421.1 |
NP_001243350.1
|
9, 10 |
1137 |
|
NM_001256422.1 |
NP_001243351.1
|
9, 10 |
1141 |
|
NM_032717.4 |
NP_116106.2
|
8, 9 |
1224 |
|
XM_011532383.1 |
XP_011530685.1
|
7, 8 |
1030 |
|
XM_011532384.2 |
XP_011530686.1
|
9, 10 |
3225 |
|
XM_017008780.1 |
XP_016864269.1
|
9, 10 |
1293 |
|
XM_017008781.1 |
XP_016864270.1
|
9, 10 |
1300 |
|
GenBank mRNA |
AB075872.1 |
- |
5, 6 |
422 |
|
AK055749.1 |
- |
9, 10 |
1136 |
|
AK223543.1 |
- |
9, 10 |
1108 |
|
AY358100.1 |
- |
8, 9 |
1041 |
|
BC073136.1 |
- |
3, 4 |
200 |
|
BC090956.1 |
- |
8, 9 |
1130 |
30387839 |
DQ324782.1 |
- |
8, 9 |
921 |
|
DQ345298.1 |
- |
8, 9 |
1221 |
|
View Full Product Details