Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_032836.2 | NP_116225.2 | 2 | 265 | |
XM_005259352.4 | XP_005259409.1 | 2 | 1290 | ||
GenBank mRNA | AK027674.1 | - | 2 | 265 | |
AK160385.1 | - | 1 | 4278 | ||
BC020368.1 | - | 1 | 567 | ||
BC132777.1 | - | 1 | 226 | 40146799 | |
BC136512.1 | - | 1 | 224 | 9020502 |