Gene Transcripts

Gene Symbol
RELT
Entrez Gene ID
84957
Gene Name
RELT tumor necrosis factor receptor
Gene Aliases
TNFRSF19L, TRLT
Chromosome Location
Chr. 11: 73376360 - 73397474 on Build GRCh38
UniGene ID
Hs.533720
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032871.3 NP_116260.2 10 1301
NM_152222.1 NP_689408.1 10 1239
XM_005274356.2 XP_005274413.1 5 1402
XM_006718714.2 XP_006718777.1 8 866
XM_011545305.1 XP_011543607.1 9 1124
GenBank mRNA AF319553.1 - 10 1301
AK027899.1 - 10 1239
AK057036.1 - 4 1043
AK074128.1 - 2 2974
AK310103.1 - 7 1107
BC017279.2 - 10 1164 4873951
BC051810.1 - 10 1191 5216988

View Full Product Details