Gene Transcripts

Gene Symbol
KRTAP4-1
Entrez Gene ID
85285
Gene Name
keratin associated protein 4-1
Gene Aliases
KAP4.1, KAP4.10, KRTAP4-10, KRTAP4.10
Chromosome Location
Chr. 17: 41184100 - 41184895 on Build GRCh38
UniGene ID
Hs.437079
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_033060.2 NP_149049.1 2 683
GenBank mRNA AJ406942.1 - 2 683
BC065372.1 - 3 1180

View Full Product Details